Learn More
Chemglass Life Sciences Cap, GL-45, Blue, Vented, Non-Sterile, Polypropylene
SDP
Catalog No. NC2823458
Encompass_Preferred
Polypropylene vented cap has a 0. 22um (PTFE) hydrophobic membrane to allow gas exchange while preventing contamination.They have a liner-less plug seal design for efficient sealing.They are autoclavable at 121 degreesC.
Catalog No. NC2823458 Supplier Chemglass Life Sciences Supplier No. CLS1484B45
Polypropylene vented cap has a 0. 22um (PTFE) hydrophobic membrane to allow gas exchange while preventing contamination.They have a liner-less plug seal design for efficient sealing.They are autoclavable at 121 degreesC.