Learn More
Chemglass Life Sciences Cap, GL-45, Blue, Vented, Non-Sterile, Polypropylene
SDP
Supplier:  Chemglass Life Sciences CLS148412
Encompass_Preferred
Polypropylene vented cap has a 0. 20um (ePTFE) hydrophobic membrane combined with larger vent holes to increase the rates of gas exchange while preventing contamination.They have a liner-less plug seal design for efficient sealing and are autoclavable at 121 degreesC.
Catalog No. NC0197973