Learn More
Chemglass Life Sciences Cap, GL-45, Clear, Solid, PFA 6515 TZ with Lip Seal
SDP
Catalog No. 5000083192
Encompass_Preferred
These premium clear caps are made from fluoropolymer (PFA 6515 TZ) with lip molded seals.Material PFA 6515 TZ is compliant to USP Class VI.Premium cap compound is formulated without color dye to avoid risk of leaching.GL-45 caps can be used in temperatures from -196 degrees to 260 degreesC.
Catalog No. 50-000-83192 Supplier Chemglass Life Sciences Supplier No. CLS148020
These premium clear caps are made from fluoropolymer (PFA 6515 TZ) with lip molded seals.Material PFA 6515 TZ is compliant to USP Class VI.Premium cap compound is formulated without color dye to avoid risk of leaching.GL-45 caps can be used in temperatures from -196 degrees to 260 degreesC.