Learn More
Chemglass Life Sciences Cap, GL-45, Clear, Solid, TpCh260 with PTFE-Coated Silicone Seal
SDP
Catalog No. 5000083227
Encompass_Preferred
These premium clear caps are made from TpCh260 (similar to PFA) with PTFE coated silicone seals.Material TpCh260 is compliant to USP Class VI.Premium cap compound is formulated without color dye to avoid risk of leaching.Premium caps can be used in temperatures from -196 degrees to 260 degreesC.
Catalog No. 50-000-83227 Supplier Chemglass Life Sciences Supplier No. CLS148019
These premium clear caps are made from TpCh260 (similar to PFA) with PTFE coated silicone seals.Material TpCh260 is compliant to USP Class VI.Premium cap compound is formulated without color dye to avoid risk of leaching.Premium caps can be used in temperatures from -196 degrees to 260 degreesC.