Learn More
Chemglass Life Sciences Cap, GL-45, Green, Solid, Polypropylene
SDP
Catalog No. 5000083215
Encompass_Preferred
Solid polypropylene caps with a molded in lip seal.All color caps can be autoclaved in steam to 140 degreesC.
Catalog No. 50-000-83215 Supplier Chemglass Life Sciences Supplier No. CLS148015
Solid polypropylene caps with a molded in lip seal.All color caps can be autoclaved in steam to 140 degreesC.