Learn More
Chemglass Life Sciences Cap, GL-45, Yellow, Open Top, (1-5/16" Dia Hole) Polypropylene
SDP
Catalog No. 501215140
Encompass
Open top polypropylene caps for use with port assemblies.Caps can be autoclaved in steam to 140 degreesC.
Catalog No. 50-121-5140 Supplier Chemglass Life Sciences Supplier No. CLS148123
Open top polypropylene caps for use with port assemblies.Caps can be autoclaved in steam to 140 degreesC.