Learn More
Chemglass Life Sciences Cap, GL-45, Yellow, Solid, Polypropylene
SDP
Catalog No. 5000083167
Encompass_Preferred
Solid polypropylene caps with a molded in lip seal.All color caps can be autoclaved in steam to 140 degreesC.
Catalog No. 50-000-83167 Supplier Chemglass Life Sciences Supplier No. CLS148018
Solid polypropylene caps with a molded in lip seal.All color caps can be autoclaved in steam to 140 degreesC.