Learn More
Ace Glass, Inc. Cap, GL25, polypropylene, blue, cs/10
SDP
Catalog No. NC0108034
Encompass
Case of 10 blue polypropylene screw caps for use with GL25 threaded bottles. For use in temperature range of -40 to 140C. Dimensions: 33mm D x 19mm H.
Catalog No. NC0108034 Supplier Ace Glass, Inc. Supplier No. 762202
Case of 10 blue polypropylene screw caps for use with GL25 threaded bottles. For use in temperature range of -40 to 140C. Dimensions: 33mm D x 19mm H.