Learn More
Metrohm USA CAP GL45
SDP
Supplier:  Metrohm USA 61602105
Encompass
6.1602.105 Bottle attachment / GL 45 / green For direct dosing from reagent bottles with GL 45 thread.
Catalog No. NC9816793