Learn More
Ace Glass, Inc. Cap, GL45, blue polypropylene, open-top, PTFE membrane, cs/5
SDP
Catalog No. 50975172
Encompass
Blue Polypropylene GL threaded cap with center hole and welded-in PTFE membrane. Membrane is 0.2uM PTFE gas permeable, not liquid permeable for pressure equalization. Temp range to 140 degrees C. Autoclavable. Great for media sterilization and storage.
Catalog No. 50-975-172 Supplier Ace Glass, Inc. Supplier No. 762945
Blue Polypropylene GL threaded cap with center hole and welded-in PTFE membrane. Membrane is 0.2uM PTFE gas permeable, not liquid permeable for pressure equalization. Temp range to 140 degrees C. Autoclavable. Great for media sterilization and storage.