Learn More
Hunter Lab 20 mm Transmission Cell with Screw Top
SDP
Catalog No. NC0854128
Encompass
HunterLab offers sealed top cells in all standard cell path lengths for the color measurement of transparent liquids that have volatiles; are corrosive or if spills are a problem. These capped cells are the same as our regular cells with the addition of a annealed glass top; two screw tops (one for pouring, one for air purge) with black polypropylene caps and white Teflon liners. 20 mm path length x 55 mm wide x 57 mm high
Catalog No. NC0854128 Supplier Hunter Lab Supplier No. D121011890
HunterLab offers sealed top cells in all standard cell path lengths for the color measurement of transparent liquids that have volatiles; are corrosive or if spills are a problem. These capped cells are the same as our regular cells with the addition of a annealed glass top; two screw tops (one for pouring, one for air purge) with black polypropylene caps and white Teflon liners. 20 mm path length x 55 mm wide x 57 mm high