Learn More
Chemglass Life Sciences Cap, GPI 38-430, Polypropylene, Sterile, Individually Bagged (20 per case) - for 38-430 threaded laboratory bottles
SDP
Catalog No. NC2399409
Encompass
Sterile polypropylene GPI 38-430 screw cap designed to seal laboratory media, reagent, and feed bottles. Caps are manufactured from Class VI polypropylene and are gamma sterilized and individually bagged for aseptic use.
  • Material: Class VI polypropylene.
  • Thread size: 38-430 (GPI).
  • Sterility: Gamma sterilized and individually bagged.
  • Quantity: 20 caps per case.
  • Intended use: Seals media, reagent, and feed bottles with 38-430 threads.
Catalog No. NC2399409 Supplier Chemglass Life Sciences Supplier No. CLS11681038
Sterile polypropylene GPI 38-430 screw cap designed to seal laboratory media, reagent, and feed bottles. Caps are manufactured from Class VI polypropylene and are gamma sterilized and individually bagged for aseptic use.
  • Material: Class VI polypropylene.
  • Thread size: 38-430 (GPI).
  • Sterility: Gamma sterilized and individually bagged.
  • Quantity: 20 caps per case.
  • Intended use: Seals media, reagent, and feed bottles with 38-430 threads.