Learn More
Chemglass Life Sciences Cap, Green with PTFE Liner, 13-425
SDP
Catalog No. 501531842
Encompass_Preferred
Cap, Green with PTFE Liner, 13-425. These caps provide a wide range of chemical capability and temperature tolerance. The foam backed PTFE resists attack from virtually all chemicals at room temperature while providing for a leak tight seal. NOT AUTOCLAVABLE.
Catalog No. 50-153-1842 Supplier Chemglass Life Sciences Supplier No. CG89502
Cap, Green with PTFE Liner, 13-425. These caps provide a wide range of chemical capability and temperature tolerance. The foam backed PTFE resists attack from virtually all chemicals at room temperature while providing for a leak tight seal. NOT AUTOCLAVABLE.