Learn More
Microsolv Technology Corporation Cap Hs Crimp Gry Butyl Rub Std
Catalog No. 5001451855
Encompass_Preferred
Caps, Headspace, Crimp Top, with Grey Butyl Rubber Laminated PTFE Septa, fitted in Aluminum, Silver Caps with 10mm center holes. For use with 20mm Crimp Top Headspace Vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51855 Supplier Microsolv Technology Corporation Supplier No. 95025021S
Caps, Headspace, Crimp Top, with Grey Butyl Rubber Laminated PTFE Septa, fitted in Aluminum, Silver Caps with 10mm center holes. For use with 20mm Crimp Top Headspace Vials. MicroSolv Brand. 100/PK.