Learn More
Microsolv Technology Corporation Cap Hs Crimp Gry Butyl Rub
Catalog No. 5001451850
Encompass_Preferred
Caps, Headspace, Crimp Top, with Dark Grey Butyl Rubber Septa, fitted in Aluminum, Silver Caps with 10mm center holes. For use with 20mm Crimp Top Headspace Vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51850 Supplier Microsolv Technology Corporation Supplier No. 95025011S
Caps, Headspace, Crimp Top, with Dark Grey Butyl Rubber Septa, fitted in Aluminum, Silver Caps with 10mm center holes. For use with 20mm Crimp Top Headspace Vials. MicroSolv Brand. 100/PK.