Learn More
Microsolv Technology Corporation Cap Hs Crimp Mag Sil/Ptfe Blue
Catalog No. 5001451862
Encompass_Preferred
Caps, Headspace, Crimp Top, with Transparent Blue Silicone Rubber / Transparent PTFE Septa, fitted in Steel, Magnetic, Gold Caps with 8mm center holes. For use with 20mm Crimp Top Headspace Vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51862 Supplier Microsolv Technology Corporation Supplier No. 95025044D
Caps, Headspace, Crimp Top, with Transparent Blue Silicone Rubber / Transparent PTFE Septa, fitted in Steel, Magnetic, Gold Caps with 8mm center holes. For use with 20mm Crimp Top Headspace Vials. MicroSolv Brand. 100/PK.