Learn More
Microsolv Technology Corporation Cap Hs Crimp Safety Dark Grey
Catalog No. 5001451851
Encompass_Preferred
Caps, Headspace, Crimp Top, with Dark Grey Butyl Rubber Septa, fitted in Aluminum, Silver Pressure Relief Caps with 10mm center holes. For use with 20mm Crimp Top Headspace Vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51851 Supplier Microsolv Technology Corporation Supplier No. 95025012S
Caps, Headspace, Crimp Top, with Dark Grey Butyl Rubber Septa, fitted in Aluminum, Silver Pressure Relief Caps with 10mm center holes. For use with 20mm Crimp Top Headspace Vials. MicroSolv Brand. 100/PK.