Learn More
Microsolv Technology Corporation Cap Hs Crimp Safety Gry Butyl
Catalog No. 5001451856
Encompass_Preferred
Caps, Headspace, Crimp Top, with Grey Butyl Rubber Laminated PTFE Septa, fitted in Aluminum, Silver Pressure Relief Caps with 8mm center holes. For use with 20mm Crimp Top Headspace Vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51856 Supplier Microsolv Technology Corporation Supplier No. 95025022S
Caps, Headspace, Crimp Top, with Grey Butyl Rubber Laminated PTFE Septa, fitted in Aluminum, Silver Pressure Relief Caps with 8mm center holes. For use with 20mm Crimp Top Headspace Vials. MicroSolv Brand. 100/PK.