Learn More
Microsolv Technology Corporation Cap Hs Crimp Safety Sil/Alum 2
Catalog No. 5001451876
Encompass_Preferred
Caps, Headspace, Crimp Top, with White Silicone Rubber / Foil (125mil) Septa, fitted in Aluminum, Silver Pressure Relief Caps with 8mm center holes. For use with 20mm Crimp Top Headspace vials. BASIK Brand. 100/PK.
Catalog No. 50-014-51876 Supplier Microsolv Technology Corporation Supplier No. 95025142S
Caps, Headspace, Crimp Top, with White Silicone Rubber / Foil (125mil) Septa, fitted in Aluminum, Silver Pressure Relief Caps with 8mm center holes. For use with 20mm Crimp Top Headspace vials. BASIK Brand. 100/PK.