Learn More
Microsolv Technology Corporation Cap Hs Crimp Safety Sil/Ptfe B
Catalog No. 5001451860
Encompass_Preferred
Caps, Headspace, Crimp Top, with Translucent Blue Silicone Rubber / White PTFE Septa, fitted in Aluminum, Silver Pressure Relief Caps with 8mm center holes. For use with 20mm Crimp Top Headspace Vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51860 Supplier Microsolv Technology Corporation Supplier No. 95025042S
Caps, Headspace, Crimp Top, with Translucent Blue Silicone Rubber / White PTFE Septa, fitted in Aluminum, Silver Pressure Relief Caps with 8mm center holes. For use with 20mm Crimp Top Headspace Vials. MicroSolv Brand. 100/PK.