Learn More
Microsolv Technology Corporation Cap Hs Crimp Sil/Alum 20Mm
Catalog No. 5001451875
Encompass_Preferred
Caps, Headspace, Crimp Top, with Natural Silicone Rubber / Foil (125mil), High Purity Septa, fitted in Aluminum, Silver Caps with 10mm center holes. For use with 20mm Crimp Top Headspace Vials. BASIK Brand. 100/PK.
Catalog No. 50-014-51875 Supplier Microsolv Technology Corporation Supplier No. 95025141S
Caps, Headspace, Crimp Top, with Natural Silicone Rubber / Foil (125mil), High Purity Septa, fitted in Aluminum, Silver Caps with 10mm center holes. For use with 20mm Crimp Top Headspace Vials. BASIK Brand. 100/PK.