Learn More
Microsolv Technology Corporation Cap Hs Crimp Sil/Ptfe Blue/Whi
Catalog No. 5001451859
Encompass_Preferred
Caps, Headspace, Crimp Top, with Translucent Blue Silicone Rubber / White PTFE Septa, fitted in Aluminum, Silver Caps with 10mm center holes. For use with 20mm Crimp Top Headspace Vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51859 Supplier Microsolv Technology Corporation Supplier No. 95025041S
Caps, Headspace, Crimp Top, with Translucent Blue Silicone Rubber / White PTFE Septa, fitted in Aluminum, Silver Caps with 10mm center holes. For use with 20mm Crimp Top Headspace Vials. MicroSolv Brand. 100/PK.