Learn More
Microsolv Technology Corporation Cap Hs Crimp Sil/Ptfe Slvr W/T
Catalog No. 5001451865
Encompass_Preferred
Caps, Headspace, Crimp Top, with White Silicone Rubber / Natural Beige PTFE Septa, fitted in Aluminum, Silver Caps with 10mm center holes. For use with 20mm Crimp Top Headspace Vials. BASIK Brand. 100/PK.
Catalog No. 50-014-51865 Supplier Microsolv Technology Corporation Supplier No. 95025071S
Caps, Headspace, Crimp Top, with White Silicone Rubber / Natural Beige PTFE Septa, fitted in Aluminum, Silver Caps with 10mm center holes. For use with 20mm Crimp Top Headspace Vials. BASIK Brand. 100/PK.