Learn More
Microsolv Technology Corporation Cap Hs Safecrimpsil/Ptfe White
Catalog No. 5001451867
Encompass_Preferred
Caps, Headspace, Crimp Top, with White Silicone Rubber / Natural Transparent PTFE, High Purity Septa, fitted in Aluminum, Silver Pressure Relief Caps with 8mm center holes. For use with 20mm Crimp Top Headspace Vials. BASIK Brand. 100/PK.
Catalog No. 50-014-51867 Supplier Microsolv Technology Corporation Supplier No. 95025082S
Caps, Headspace, Crimp Top, with White Silicone Rubber / Natural Transparent PTFE, High Purity Septa, fitted in Aluminum, Silver Pressure Relief Caps with 8mm center holes. For use with 20mm Crimp Top Headspace Vials. BASIK Brand. 100/PK.