Learn More
Microsolv Technology Corporation Cap Hs Scrw 55S Butyl/Ptfe Cd
Supplier:  Microsolv Technology Corporation 9502055SCDK
Encompass_Preferred
Caps, Headspace, Screw Top, Closed, with Reddish Butyl Rubber / Grey PTFE Liner, fitted in Steel, Magnetic, Silver Caps with 8mm center holes. For use with 18mm Precision Thread Headspace Vials. MicroSolv Brand. 1000/CS.
Catalog No. 50-014-51843
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.