Learn More
Microsolv Technology Corporation Cap Hs Scrw Mag 45S Sil/Ptfe
Catalog No. 5001451840
Encompass_Preferred
Caps, Headspace, Screw Top, with White Silicone Rubber / Red PTFE Septa, fitted in Steel, Magnetic, Silver Caps with 8mm center holes. For use with 18mm Precision Thread Headspace Vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51840 Supplier Microsolv Technology Corporation Supplier No. 9502045SW
Caps, Headspace, Screw Top, with White Silicone Rubber / Red PTFE Septa, fitted in Steel, Magnetic, Silver Caps with 8mm center holes. For use with 18mm Precision Thread Headspace Vials. MicroSolv Brand. 100/PK.