Learn More
Microsolv Technology Corporation Cap Hs Scrw Mag 55S Sil/Ptfe
Catalog No. 5001451845
Encompass_Preferred
Caps, Headspace, Screw Top, with White Silicone Rubber / Blue PTFE Temperature Resistant Septa, fitted in Steel, Magnetic, Silver Caps with 8mm center holes. For use with 18mm Precision Thread Headspace Vials. MicroSolv Brand. 1000/CS.
Catalog No. 50-014-51845 Supplier Microsolv Technology Corporation Supplier No. 9502055SWK
Caps, Headspace, Screw Top, with White Silicone Rubber / Blue PTFE Temperature Resistant Septa, fitted in Steel, Magnetic, Silver Caps with 8mm center holes. For use with 18mm Precision Thread Headspace Vials. MicroSolv Brand. 1000/CS.