Learn More
Microsolv Technology Corporation Cap Hscrimpsil/Ptfe White/Whit
Catalog No. 5001451866
Encompass_Preferred
Caps, Headspace, Crimp Top, with White Silicone Rubber / Natural Translucent PTFE, High Purity Septa, fitted in Aluminum, Silver Caps with 10mm center holes. For use with 20mm Crimp Top Headspace Vials. BASIK Brand. 100/PK.
Catalog No. 50-014-51866 Supplier Microsolv Technology Corporation Supplier No. 95025081S
Caps, Headspace, Crimp Top, with White Silicone Rubber / Natural Translucent PTFE, High Purity Septa, fitted in Aluminum, Silver Caps with 10mm center holes. For use with 20mm Crimp Top Headspace Vials. BASIK Brand. 100/PK.