Learn More
Research Products International Corp Cap Inserts, 0.5-2.0ml, Gray, 100/pk
SDP
Catalog No. 501978659
Encompass
The Cap Insert is used in 0.5-2.0ml CryoStore Vials to color code according to your specific needs. It is inserted on top of the closure and has a write-on frosted area for sample identification.
Catalog No. 50-197-8659 Supplier Research Products International Corp Supplier No. 212435GY
The Cap Insert is used in 0.5-2.0ml CryoStore Vials to color code according to your specific needs. It is inserted on top of the closure and has a write-on frosted area for sample identification.