Learn More
Waters Corp Cap-mat 96-well 7 mm Round Plug Pre-slit Silicone/PTFE, 5/pk
Catalog No. 50466283 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Pre-slit PTFE / Silicone Cap Mat for 96-well plates with 7 mm round wells and 1 mL glass inserts.
Catalog No. 50-466-283 Supplier Waters Corp Supplier No. 186006332
Pre-slit PTFE / Silicone Cap Mat for 96-well plates with 7 mm round wells and 1 mL glass inserts.