Learn More
Waters Corp Waters Cap Mat, 96-well, Square Plug, Pre-slit Silicone/PTFE
Catalog No. 50466286 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This pre-slit PTFE/Silicone Cap Mat for 96-well plates with square wells and 700 μL glass inserts offers protection during storage, preventing contamination and evaporation to ensure analyte purity. It is compatible with thick and/or blunt HPLC needles.
  • Protects all 96 wells during storage, preventing contamination and evaporation.
  • Compatible with thick/blunt HPLC needles; eliminates coring and offers good resealing for multiple injections.
  • Designed for 96-well plates with square wells and 700 μL glass inserts.
  • Pre-slit Silicone/PTFE ensures adequate venting and excellent sample-draw reproducibility.
  • Working temperature range: -40°C to 200°C.
  • Recommended for autosamplers to promote proper venting and accurate sample draw.
Catalog No. 50-466-286 Supplier Waters Corp Supplier No. 186006335
This pre-slit PTFE/Silicone Cap Mat for 96-well plates with square wells and 700 μL glass inserts offers protection during storage, preventing contamination and evaporation to ensure analyte purity. It is compatible with thick and/or blunt HPLC needles.
  • Protects all 96 wells during storage, preventing contamination and evaporation.
  • Compatible with thick/blunt HPLC needles; eliminates coring and offers good resealing for multiple injections.
  • Designed for 96-well plates with square wells and 700 μL glass inserts.
  • Pre-slit Silicone/PTFE ensures adequate venting and excellent sample-draw reproducibility.
  • Working temperature range: -40°C to 200°C.
  • Recommended for autosamplers to promote proper venting and accurate sample draw.