Learn More
Thomas Scientific Cap Mat, EVA, Sterile, Round Nap Shape for 1mL MASTERBLOCK & 96W PP Plates, Natural
SDP
Catalog No. 50320720
Encompass_Preferred
Free of detectable DNase, RNase, human DNA. Non-Pyrogenic. Resistant to DMSO. Can be used in a temperature range between -20°C and + 60°C. The 96 well F-, U-, and µClear-bottom polystyrene microplates may also be sealed (except 96 well V-bottom polystyrene microplates). Not pierceable.
Catalog No. 50-320-720 Supplier Thomas Scientific Supplier No. 1176P75
Free of detectable DNase, RNase, human DNA. Non-Pyrogenic. Resistant to DMSO. Can be used in a temperature range between -20°C and + 60°C. The 96 well F-, U-, and µClear-bottom polystyrene microplates may also be sealed (except 96 well V-bottom polystyrene microplates). Not pierceable.