Learn More
Waters Corp Waters Cap-mat, Preslit PTFE/Silicone, 96-Well, 5-pack
Catalog No. 50819164 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This cap mat features preslit PTFE/silicone septa, designed for use with 96- and 384-well sample and collection plates. It facilitates easy needle penetration and is intended for single-use applications in various laboratory settings.
  • Pre-slit PTFE and silicone septa for easy needle penetration.
  • Designed for single-use.
  • Compatible with most standard vials and containers.
  • Compatible with most automated liquid handling systems.
  • Offers square wells.
Catalog No. 50-819-164 Supplier Waters Corp Supplier No. 186000857
This cap mat features preslit PTFE/silicone septa, designed for use with 96- and 384-well sample and collection plates. It facilitates easy needle penetration and is intended for single-use applications in various laboratory settings.
  • Pre-slit PTFE and silicone septa for easy needle penetration.
  • Designed for single-use.
  • Compatible with most standard vials and containers.
  • Compatible with most automated liquid handling systems.
  • Offers square wells.