Learn More
Brookfield Engineer Labs High Torque CAP2L Mineral Oil Fluid, Low Temp 25 ° C
Catalog No. NC1850168
Encompass
General purpose oil fluids are best suited for those instruments using Cone/Plate Viscometers/Rheometers. Brookfield Viscosity Standards are accurate to ±1% of the stated viscosity and are certified by methods traceable to the United States National Institute of Standards and Technology (NIST). The selection of one or two fluids will normally provide sufficient measurement points to verify calibration of your instrument.
Catalog No. NC1850168 Supplier Brookfield Engineer Labs Supplier No. CAP2L
General purpose oil fluids are best suited for those instruments using Cone/Plate Viscometers/Rheometers. Brookfield Viscosity Standards are accurate to ±1% of the stated viscosity and are certified by methods traceable to the United States National Institute of Standards and Technology (NIST). The selection of one or two fluids will normally provide sufficient measurement points to verify calibration of your instrument.