Learn More
CELLTREAT Scientific Products CAP ONLY, 14mL Culture Tube Dual Cap, PE, Non-Sterile, 1000/cs
SDP
Catalog No. NC3433463
Encompass_Preferred
"Dual caps allow for either vented or sealed positions14mL tubes feature molded graduationsPolystyrene tubes are optically clear to visualize culturesPolypropylene tubes offer better chemical resistance"
Catalog No. NC3433463 Supplier CELLTREAT Scientific Products Supplier No. 230444
"Dual caps allow for either vented or sealed positions14mL tubes feature molded graduationsPolystyrene tubes are optically clear to visualize culturesPolypropylene tubes offer better chemical resistance"