Learn More
Ace Glass, Inc. CAP ONLY CAP ONLY
SDP
Supplier:  Ace Glass, Inc. 541507
Encompass_Preferred
Cap with 5/12 standard taper joint, for use with 10mL and 50mL Specific Gravity bottles.
Catalog No. 50-971-877