Learn More
CELLTREAT Scientific Products CAP ONLY, Screw Top Micro Tube Cap, O-Ring, Translucent, Clear, Sterile, 1000/cs
SDP
Catalog No. NC3433472
Encompass_Preferred
"Translucent and Opaque Cap O-rings ensure leak-proof seal, and are a highly visible contrasting colorAll caps are compatible with all CELLTREAT Screw Top Micro Tubes; mix and match"
Catalog No. NC3433472 Supplier CELLTREAT Scientific Products Supplier No. 230842CS
"Translucent and Opaque Cap O-rings ensure leak-proof seal, and are a highly visible contrasting colorAll caps are compatible with all CELLTREAT Screw Top Micro Tubes; mix and match"