Learn More
Chemglass Life Sciences Cap, Green, Open Top, 13-425, with 0.60" PP/PTFE/Silicone SURE-LINK Septa
SDP
Catalog No. 50143880
Encompass_Preferred
Cap, Green, Open Top, 13-425, with 0.60" PP/PTFE/Silicone SURE-LINK Septa. These caps, made of polypropylene, are ideal for use with volatile solvents, sensitive compounds, and biological media. The SURE-LINK process welds each cap with a Polypropylene/PTFE/Silicone liner bonded to the cap. This process allows the cap to maintain seal through multiple autoclaving operations or cleaning procedures without the use of adhesives. Caps are ideal for storage of samples for prolonged periods of time. They are autoclavable at 121¿C.
Catalog No. 50-143-880 Supplier Chemglass Life Sciences Supplier No. CG491015
Cap, Green, Open Top, 13-425, with 0.60" PP/PTFE/Silicone SURE-LINK Septa. These caps, made of polypropylene, are ideal for use with volatile solvents, sensitive compounds, and biological media. The SURE-LINK process welds each cap with a Polypropylene/PTFE/Silicone liner bonded to the cap. This process allows the cap to maintain seal through multiple autoclaving operations or cleaning procedures without the use of adhesives. Caps are ideal for storage of samples for prolonged periods of time. They are autoclavable at 121¿C.