Learn More
Chemglass Life Sciences Cap, Open Hole, Black Polypropylene, GPI 18-400 with Gray PTFE/Silicone
SDP
Catalog No. 501532563
Encompass_Preferred
Cap, Open Hole, Black Polypropylene, GPI 18-400 with Gray PTFE/Silicone. Designed for use with Shimadzu, Spectra-Physics, Varian and other autosamplers. The Top Seal closure with a 0.010 molded septa allows for easy needle penetration and is inert to most solvents. The one-piece design saves time by eliminating liners in the closures.
Catalog No. 50-153-2563 Supplier Chemglass Life Sciences Supplier No. CV37300018
Cap, Open Hole, Black Polypropylene, GPI 18-400 with Gray PTFE/Silicone. Designed for use with Shimadzu, Spectra-Physics, Varian and other autosamplers. The Top Seal closure with a 0.010 molded septa allows for easy needle penetration and is inert to most solvents. The one-piece design saves time by eliminating liners in the closures.