Learn More
Chemglass Life Sciences Closure Only, GPI 13-425, Green, Open Hole, Polypropylene
SDP
Catalog No. 501532564
Encompass_Preferred
Closure Only, GPI 13-425, Green, Open Hole, Polypropylene. designed for use with Shimadzu, Spectra-Physics, Varian and other autosamplers. The Top Seal closure with a 0.010 molded septa allows for easy needle penetration and is inert to most solvents. The one-piece design saves time by eliminating liners in the closures.
Catalog No. 50-153-2564 Supplier Chemglass Life Sciences Supplier No. CV3730G013
Closure Only, GPI 13-425, Green, Open Hole, Polypropylene. designed for use with Shimadzu, Spectra-Physics, Varian and other autosamplers. The Top Seal closure with a 0.010 molded septa allows for easy needle penetration and is inert to most solvents. The one-piece design saves time by eliminating liners in the closures.