Learn More
Chemglass Life Sciences Closure Only, GPI 13-425, Red, Open Hole, Polypropylene
SDP
Catalog No. 501532565
Encompass_Preferred
Closure Only, GPI 13-425, Red, Open Hole, Polypropylene. Used with Viton O-Rings and threaded outer joints for making grease-free, vacuum tight seals on 14/10 MINUM-WARE glassware components. The 8-425 size is used on the sidearm of Hickman still head and MW-81 GC collection vial, while the 13-425 is used on the MW-56 two neck flask sidearm.
Catalog No. 50-153-2565 Supplier Chemglass Life Sciences Supplier No. CV3730R013
Add to Cart
Add to Cart
Closure Only, GPI 13-425, Red, Open Hole, Polypropylene. Used with Viton O-Rings and threaded outer joints for making grease-free, vacuum tight seals on 14/10 MINUM-WARE glassware components. The 8-425 size is used on the sidearm of Hickman still head and MW-81 GC collection vial, while the 13-425 is used on the MW-56 two neck flask sidearm.