Learn More
Chemglass Life Sciences Cap, GL-45, Schott Blue, Solid, Polypropylene
SDP
Catalog No. 50143798
Encompass_Preferred
Cap, GL-45, Schott Blue, Solid, Polypropylene. Solid polypropylene caps with a molded in lip seal. All color caps can be autoclaved in steam to 140°C.
Catalog No. 50-143-798 Supplier Chemglass Life Sciences Supplier No. CLS148013SC
Cap, GL-45, Schott Blue, Solid, Polypropylene. Solid polypropylene caps with a molded in lip seal. All color caps can be autoclaved in steam to 140°C.