Learn More
Chemglass Life Sciences Cap, GC Chem Snap, Polypropylene, with PTFE/Silicone/PTFE Septum
SDP
Catalog No. 501532560
Encompass_Preferred
Cap, GC Chem Snap, Polypropylene, with PTFE/Silicone/PTFE Septum. GC Chem Snap Caps were developed to handle the volatile samples, providing a liquid airtight seal with the vial. Simply snap on the cap and an evaporation-proof seal of a crimped cap is achieved.
Catalog No. 50-153-2560 Supplier Chemglass Life Sciences Supplier No. CV36150011
Cap, GC Chem Snap, Polypropylene, with PTFE/Silicone/PTFE Septum. GC Chem Snap Caps were developed to handle the volatile samples, providing a liquid airtight seal with the vial. Simply snap on the cap and an evaporation-proof seal of a crimped cap is achieved.