Learn More
Chemglass Life Sciences Cap, Chem Snap, 11mm, Clear, PTFE/Silicone with Star Slit
SDP
Catalog No. 501532562
Encompass_Preferred
Cap, Chem Snap, 11mm, Clear, PTFE/Silicone with Star Slit. Chem Snap Caps may be snapped on and off by hand which eliminates the need for metal crimping and decapping tools, thus reducing potential hazards due to sharp aluminum edges. Large opening provides greater target area for needle penetration. Snap caps are color coded for easy sample identification.
Catalog No. 50-153-2562 Supplier Chemglass Life Sciences Supplier No. CV3630S011
Cap, Chem Snap, 11mm, Clear, PTFE/Silicone with Star Slit. Chem Snap Caps may be snapped on and off by hand which eliminates the need for metal crimping and decapping tools, thus reducing potential hazards due to sharp aluminum edges. Large opening provides greater target area for needle penetration. Snap caps are color coded for easy sample identification.