Learn More
Chemglass Life Sciences Cap, GC Chem Snap, Polypropylene, with PTFE/Silicone Septum
SDP
Catalog No. 501532559
Encompass_Preferred
Cap, GC Chem Snap, Polypropylene, with PTFE/Silicone Septum. GC Chem Snap Caps were developed to handle the volatile samples, providing a liquid airtight seal with the vial. Simply snap on the cap and an evaporation-proof seal of a crimped cap is achieved.
Catalog No. 50-153-2559 Supplier Chemglass Life Sciences Supplier No. CV35950011
Cap, GC Chem Snap, Polypropylene, with PTFE/Silicone Septum. GC Chem Snap Caps were developed to handle the volatile samples, providing a liquid airtight seal with the vial. Simply snap on the cap and an evaporation-proof seal of a crimped cap is achieved.