Learn More
Ace Glass, Inc. Cap, PTFE lined, 24-400, fits 8 oz bottles, package/6
SDP
Catalog No. 50983362
Encompass
Black plastic screw caps with PTFE liners for use with ACE 5546, 5547, and 5549 8oz bottles.
Catalog No. 50-983-362 Supplier Ace Glass, Inc. Supplier No. 1248908
Black plastic screw caps with PTFE liners for use with ACE 5546, 5547, and 5549 8oz bottles.