Learn More
Chemglass Life Sciences Cap, Red, 3mm, Low Density, Polyethylene, NMR, Tube, Standard
SDP
Catalog No. CTC3LPE
Encompass_Preferred
Cap, Red, 3mm, Low Density, Polyethylene, NMR, Tube, Standard. NORELL™ Standard NMR Tube Caps are made of low density polyethylene, and available in 3mm and 10mm.
Catalog No. CTC3LPE Supplier Chemglass Life Sciences Supplier No. CTC3LPE
Cap, Red, 3mm, Low Density, Polyethylene, NMR, Tube, Standard. NORELL™ Standard NMR Tube Caps are made of low density polyethylene, and available in 3mm and 10mm.