Learn More
Trajan Scientific L-MARK Brand Cap, Screw, Black, 13-425, Solid Top, PP, With PTFE/silicone liner, 100 Pack
Catalog No. 501179667
Encompass_Preferred
L-MARK Brand, Cap, Screw, Black, 13-425, Solid Top, Polypropylene, With PTFE/White silicone liner, 100 Pack. Made in the USA.
Catalog No. 50-117-9667 Supplier Trajan Scientific Supplier No. LPP.013139098
L-MARK Brand, Cap, Screw, Black, 13-425, Solid Top, Polypropylene, With PTFE/White silicone liner, 100 Pack. Made in the USA.