Learn More
Trajan Scientific L-MARK Brand Cap Screw Bonded Black 10-425, 100 Pack
Catalog No. 501090058
Encompass_Preferred
L-MARK Brand Screw Cap, PP, Black, PTFE/Red Rubber, 10-425, Bonded, 100 Pack. Made in the USA.
Catalog No. 50-109-0058 Supplier Trajan Scientific Supplier No. LPP.010138971
L-MARK Brand Screw Cap, PP, Black, PTFE/Red Rubber, 10-425, Bonded, 100 Pack. Made in the USA.