Learn More
Microsolv Technology Corporation Cap Screw Blk Preslit Sil/Ptfe
Catalog No. 5001452175
Encompass_Preferred
Caps, Screw Top, with White Silicone Rubber / Natural PTFE Pre-Slit Septa, fitted in knurled, polypropylene, Black Caps. For use with 9-425mm thread, 12x32mm autosampler vials. BASIK Brand. 100/PK.
Catalog No. 50-014-52175 Supplier Microsolv Technology Corporation Supplier No. 9502SE35K
Caps, Screw Top, with White Silicone Rubber / Natural PTFE Pre-Slit Septa, fitted in knurled, polypropylene, Black Caps. For use with 9-425mm thread, 12x32mm autosampler vials. BASIK Brand. 100/PK.